Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CSP alpha Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142988
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Cysteine string protein alpha (CSPalpha) is a synaptic vesicle protein that contains a DNA-J domain characteristic of Hsp40 chaperones. Cysteine-string proteins are membrane-bound proteins associated primarily with nerve endings and synaptic vesicles in the brain. These proteins play an important role in presynaptic function and may be involved in calcium-dependent neurotransmitter release. CSP proteins may act as presynaptic chaperones that assist in maintaining synaptic function.
Specifications
CSP alpha | |
Polyclonal | |
Unconjugated | |
Dnajc5 | |
2610314I24Rik; AU018536; Ceroid-lipofuscinosis neuronal protein 4; CLN4; CLN4B; Csp; CSP alpha; cysteine string protein; cysteine string protein alpha; DnaJ (Hsp40) homolog, subfamily C, member 5; DnaJ heat shock protein family (Hsp40) member C5; dnaJ homolog subfamily C member 5; Dnajc5; DNAJC5A; mir-941-2; mir-941-3; mir-941-4; mir-941-5; NCL | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
80331 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9H3Z4 | |
Dnajc5 | |
Peptide with sequence C-SATETTQLTADSHPS. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction