Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585031 Supplier Invitrogen™ Supplier No. PA585031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MACNIPNQRQ RTLSTTGEAL YEILGLHKGA SNEEIKKTYR Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 88%.

DnaJ is a testis-specific heat shock protein and functions in association with DnaK(Hsp70). It is a molecular chaperone to facilitate protein folding. DnaJ plays a key role in the chaperone reaction by stimulating the ATPase activity and activating the substrate binding of Hsp70.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC5B
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJC5B
Gene Accession No. Q9UF47
Gene Alias 1700008A05Rik; beta cysteine string protein; beta-CSP; CSP-beta; Cysteine string protein beta; DnaJ (Hsp40) homolog, subfamily C, member 5 beta; DnaJ heat shock protein family (Hsp40) member C5 beta; dnaJ homolog subfamily C member 5B; dnaJ homolog subfamily C member X; Dnajc5b; RGD1562486; testicular tissue protein Li 55
Gene Symbols DNAJC5B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJC5B. Recombinant protein control fragment (Product #RP-108679).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85479
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.