Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501731533 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1533 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1533 Supplier Proteintech Group Inc Supplier No. 173641AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DnaJ is a testis-specific heat shock protein and functions in association with DnaK(Hsp70). It is a molecular chaperone to facilitate protein folding. DnaJ plays a key role in the chaperone reaction by stimulating the ATPase activity and activating the substrate binding of Hsp70.

Specifications

Antigen DNAJC5B
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DNAJC5B
Gene Accession No. D3ZD82, Q9CQ94, Q9UF47
Gene Alias Beta CSP, Beta cysteine string protein, CSP beta, DNAJC5B
Gene Symbols Dnajc5b
Host Species Rabbit
Immunogen DNAJC5B Fusion Protein Ag10744
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499579, 66326, 85479
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.