Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC5G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116261
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116261 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116261 Supplier Invitrogen™ Supplier No. PA5116261
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNAJC5G Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC5G
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DNAJC5G
Gene Accession No. Q8N7S2
Gene Alias 4922503E23Rik; CSP-gamma; Cysteine string protein-gamma; DNAJ; DnaJ (Hsp40) homolog, subfamily C, member 5 gamma; DnaJ heat shock protein family (Hsp40) member C5 gamma; dnaJ homolog subfamily C member 5G; Dnajc5g; FLJ40417; gamma cysteine string protein; gamma-CSP; gamma-cysteine string protein
Gene Symbols DNAJC5G
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 88-115 amino acids from the Central region of human DNAJC5G.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285126
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.