Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNAJC5G Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537997
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

Specifications
DNAJC5G | |
Polyclonal | |
Unconjugated | |
DNAJC5G | |
4922503E23Rik; CSP-gamma; Cysteine string protein-gamma; DNAJ; DnaJ (Hsp40) homolog, subfamily C, member 5 gamma; DnaJ heat shock protein family (Hsp40) member C5 gamma; dnaJ homolog subfamily C member 5G; Dnajc5g; FLJ40417; gamma cysteine string protein; gamma-CSP; gamma-cysteine string protein | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
285126 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8N7S2 | |
DNAJC5G | |
Peptide with sequence C-DSGRKYQQNVQSQP, from the internal region near the C-terminus of human DNAJC5G (aa 157-171). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction