Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521543 Supplier Invitrogen™ Supplier No. PA521543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, mouse brain. Predicted reactivity: Mouse (98%), Rat (97%), Zebrafish (91%), Chicken (97%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DNAJC7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene DNAJC7
Gene Accession No. Q99615, Q9QYI3
Gene Alias 2010003F24Rik; 2010004G07Rik; CCRP; Cytoplasmic CAR retention protein; cytoplasmic constitutive active/androstane receptor retention protein; DJ11; DJC7; DnaJ (Hsp40) homolog, subfamily C, member 7; DnaJ heat shock protein family (Hsp40) member C7; dnaJ homolog subfamily C member 7; DNAJC7; MDj11; mTpr2; tetratricopeptide repeat domain 2; tetratricopeptide repeat protein 2; TPR repeat protein 2; TPR2; TTC2
Gene Symbols DNAJC7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DNAJC7. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56354, 7266
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.