Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115135 Supplier Thermo Scientific Supplier No. PA5115135
Add to Cart
Add to Cart

DNAJC7 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DNAJC7
Gene Accession No. Q99615, Q9QYI3
Gene Alias 2010003F24Rik; 2010004G07Rik; CCRP; Cytoplasmic CAR retention protein; cytoplasmic constitutive active/androstane receptor retention protein; DJ11; DJC7; DnaJ (Hsp40) homolog, subfamily C, member 7; DnaJ heat shock protein family (Hsp40) member C7; dnaJ homolog subfamily C member 7; DNAJC7; MDj11; mTpr2; tetratricopeptide repeat domain 2; tetratricopeptide repeat protein 2; TPR repeat protein 2; TPR2; TTC2
Gene Symbols DNAJC7
Host Species Rabbit
Immunogen A synthesized peptide derived from human DNAJC7(Accession Q99615), corresponding to amino acid residues M11-N61.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56354, 7266
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.