Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565000
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565000 Supplier Invitrogen™ Supplier No. PA565000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGSRGGWAGEM AASGESGTSG GGGSTEEAFM TFYSEVKQIE KRDSVLTSKN QIERLTRPGS SYFNLNPFEV LQIDPEVTDE EIK Highest antigen sequence identity to the following orthologs - mouse 87%, rat 87%.

Suppresses polyglutamine (polyQ) aggregation of ATXN3 in neuronal cells (PubMed:27133716). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC8
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJC8
Gene Accession No. O75937
Gene Alias 1110021D09Rik; 2010009J04Rik; AL024084; AU019262; AU044514; DnaJ (Hsp40) homolog, subfamily C, member 8; DnaJ heat shock protein family (Hsp40) member C8; dnaJ homolog subfamily C member 8; DNAJC8; HSPC315; HSPC331; SPF31; splicing protein spf31
Gene Symbols DNAJC8
Host Species Rabbit
Immunogen Recombinant Human DNAJC8. Recombinant protein control fragment (Product #RP-105763).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22826
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.