Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109647 Supplier Invitrogen™ Supplier No. PA5109647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAASGESGTS GGGGSTEEAF MTFYSEVKQI EKRDSVLTSK NQIERLTRPG SSYFNLNPFE VLQIDPEVTD EEIKKRFRQL SILVHPDKNQ DDADRAQKAF EAVDKAYKLL LDQEQKKRAL DVIQAGKEYV EHTVKERKKQ LKKEGKPTIV EEDDPELFKQ AVYKQTMKLF AELEIKRKER EAKEMHERKR QREEEIEAQE KAKREREWQK NFEESRDGRV DSWRNFQANT KGKKEKKNRT FLRPPKVKME QRE.

Suppresses polyglutamine (polyQ) aggregation of ATXN3 in neuronal cells (PubMed:27133716). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DNAJC8
Gene Accession No. Q6NZB0
Gene Alias 1110021D09Rik; 2010009J04Rik; AL024084; AU019262; AU044514; DnaJ (Hsp40) homolog, subfamily C, member 8; DnaJ heat shock protein family (Hsp40) member C8; dnaJ homolog subfamily C member 8; DNAJC8; HSPC315; HSPC331; SPF31; splicing protein spf31
Gene Symbols DNAJC8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-253 of human DNAJC8 (NP_055095.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68598
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.