Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109648 Supplier Invitrogen™ Supplier No. PA5109648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGLLDLCEEV FGTADLYRVL GVRREASDGE VRRGYHKVSL QVHPDRVGEG DKEDATRRFQ ILGKVYSVLS DREQRAVYDE QGTVDEDSPV LTQDRDWEAY WRLLFKKISL EDIQAFEKTY KGSEEELADI KQAYLDFKGD MDQIMESVLC VQYTEEPRIR NIIQQAIDAG EVPSYNAFVK ESKQKMNARK RRAQEEAKEA EMSRKELGLD EGVDSLKAAI QSRQKDRQKE MDNFLAQMEA KYCKSSKGGG KKSALKKEKK.

DjC9 (DnaJ subfamily C member 9) belongs to the Hsp40 (heat shock protein 40) family of proteins that function as co-chaperones to Hsp70.Hsp40 proteins are classified into 3 main subfamilies (A, B, and C). The families are defined by the presence of particular domains which include a highly conserved alpha-helical N-terminal domain termed the J-domain, a glycine/phenylalanine-rich region, a cysteine-rich region, and a C-terminal beta-sheet containing domain. DjC9 is part of subfamily C that bears only the J-domain. DjC9 has been shown to interact with HSP70 through its J domain and activate its ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC9
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DNAJC9
Gene Accession No. Q8WXX5
Gene Alias 5330419I01Rik; AU020082; DnaJ (Hsp40) homolog, subfamily C, member 9; DnaJ heat shock protein family (Hsp40) member C9; dnaJ homolog subfamily C member 9; DnaJ protein SB73; Dnajc9; DnaJ-like factor; HDJC9; JDD1; SB73
Gene Symbols DNAJC9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DNAJC9 (NP_056005.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23234
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.