Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJC9 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | DNAJC9 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4912
![]() ![]() |
Proteintech Group Inc
254441AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
DjC9 (DnaJ subfamily C member 9) belongs to the Hsp40 (heat shock protein 40) family of proteins that function as co-chaperones to Hsp70.Hsp40 proteins are classified into 3 main subfamilies (A, B, and C). The families are defined by the presence of particular domains which include a highly conserved alpha-helical N-terminal domain termed the J-domain, a glycine/phenylalanine-rich region, a cysteine-rich region, and a C-terminal beta-sheet containing domain. DjC9 is part of subfamily C that bears only the J-domain. DjC9 has been shown to interact with HSP70 through its J domain and activate its ATPase activity.Specifications
DNAJC9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8WXX5 | |
23234 | |
DNAJC9 Fusion Protein Ag22123 | |
Primary | |
-20°C | |
DNAJC9 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DnaJ protein SB73, DNAJC9, HDJC9, JDD1, KIAA0974, SB73 | |
DNAJC9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title