Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAL1 Antibody (29E4), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242657MFV610
Description
DNAL1 Monoclonal antibody specifically detects DNAL1 in Human samples. It is validated for Flow CytometrySpecifications
DNAL1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
290000 | |
Flow Cytometry | |
C14orf168MGC12435,1700010H15RiK, chromosome 14 open reading frame 168, dynein light chain 1, axonemal, dynein, axonemal, light chain 1 | |
Recombinant human DNAL1 (1-190aa) purified from E. coli | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Neurodegeneration, Neuroscience, Signal Transduction | |
83544 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction