Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588653 Supplier Invitrogen™ Supplier No. PA588653
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DASLSMLANC EKLSLSTNCI EKIANLNGLK NLRILSLGRN NIKNLNGLEA VGDTLEELWI SYNFIEKLKG IHIMKKLKIL YMSNNLVKDW AEFVKLAELP CLEDLVFVGN PLEEKHSAEN NWIEEATKRV PKLKKLDGTP VIKGDEEEDN; Positive Samples: Raji, HeLa, HepG2, U-251, Mouse brain, Rat brain, Rat testis.

DNAL1 was identified as a potential candidate gene for primary ciliary dyskinesia (PCD), a genetically heterologous disorder characterized by chronic infections of the upper and lower airways that often leads to permanent lung damage, randomization of left/right body symmetry, and reduced fertility. DNAL1 is reported to be expressed solely in tissues carrying motile cilia for flagella and interacts with DNAH5, a protein that when mutated has been shown to result in PCD. It has been suggested that DNAL1 serves a regulatory function for DNAH5 activity in outer dynein arms of sperm flagella, respiratory cilia, and ependymal cilia. DNAL1 has also been recently identified as an HIV dependency factor (HDF), suggesting that DNAL1 may be an important drug target in HIV treatment. At least two isoforms of DNAL1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Dnal1
Gene Accession No. Q05A62, Q4LDG9
Gene Alias 1700010H15Rik; AW121714; axonemal dynein light chain 1; C14orf168; CILD16; DNAL1; Dnalc1; dynein axonemal light chain 1; dynein light chain 1, axonemal; dynein, axonemal, light chain 1; E330027P08Rik; LC1
Gene Symbols Dnal1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 41-190 of human DNAL1 (NP_1136152).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105000, 685664, 83544
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.