Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109649 Supplier Thermo Scientific Supplier No. PA5109649
Add to Cart
Add to Cart

DNAL1 Polyclonal Antibody

DNAL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAL1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Dnal1
Gene Accession No. Q05A62, Q4LDG9
Gene Alias 1700010H15Rik; AW121714; axonemal dynein light chain 1; C14orf168; CILD16; DNAL1; Dnalc1; dynein axonemal light chain 1; dynein light chain 1, axonemal; dynein, axonemal, light chain 1; E330027P08Rik; LC1
Gene Symbols Dnal1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 41-190 of human DNAL1 (NP_113615.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105000, 685664, 83544
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.