Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588420 Supplier Invitrogen™ Supplier No. PA588420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGETEGKKDE ADYKRLQTFP LVRHSDMPEE MRVETMELCV TACEKFSNNN ESAAKMIKET MDKKFGSSWH VVIGEGFGFE ITHEVKNLLY LYFGGTLAVC VWKCS; Positive Samples: Mouse testis, Rat testis; Cellular Location: Cytoplasm, cilium axoneme, cytoskeleton.

Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules. Dynein has ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAL4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DNAL4
Gene Accession No. Q9DCM4
Gene Alias D15Ertd424e; DNAL4; Dnalc4; dynein axonemal light chain 4; dynein light chain 4, axonemal; dynein light chain, outer arm 4; dynein, axon, light chain 4; dynein, axonemal, light chain 4; dynein, axonemal, light polypeptide 4; MRMV3; outer arm dynein light chain 4; PIG27; proliferation-inducing gene 27; proliferation-inducing protein 27
Gene Symbols DNAL4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human DNAL4 (NP_0057311).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300078, 54152
Target Species Mouse, Rat, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.