Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNALI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555816 Supplier Invitrogen™ Supplier No. PA555816
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CWVATLASAM IPPADSLLKY DTPVLVSRNT EKRSPKARLL KVSPQQPGPS GSAPQPPKTK LPSTPCVPDP TKQ Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 78%.

This gene is the human homolog of the Chlamydomonas inner dynein arm gene, p28. The precise function of this gene is not known, however, it is a potential candidate for immotile cilia syndrome (ICS). Ultrastructural defects of the inner dynein arms are seen in patients with ICS. Immotile mutant strains of Chlamydomonas, a biflagellated algae, exhibit similar defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNALI1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNALI1
Gene Accession No. O14645
Gene Alias 1700023A09Rik; AW049135; axonemal dynein light chain (hp28) homolog; axonemal dynein light intermediate polypeptide 1; axonemal dynein light intermediate polypeptide 1; LOW QUALITY PROTEIN: axonemal dynein light intermediate polypeptide 1; dJ423B22.5; dJ423B22.5 (axonemal dynein light chain (hp28)); Dnali1; dynein axonemal light intermediate chain 1; dynein, axonemal, light intermediate chain 1; dynein, axonemal, light intermediate polypeptide 1; hp28; Inner dynein arm light chain, axonemal; inner dynein arm, homolog of clamydomonas; LOW QUALITY PROTEIN: axonemal dynein light intermediate polypeptide 1; P28
Gene Symbols DNALI1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNALI1. Recombinant protein control fragment (Product #RP-94440).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.