Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAM-1/CD226 Antibody (102511) [DyLight 650], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB666W
Description
DNAM-1/CD226 Monoclonal antibody specifically detects DNAM-1/CD226 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOFSpecifications
DNAM-1/CD226 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Neutralization, CyTOF | |
102511 | |
Western Blot, Flow Cytometry, Neutralization, CyTOF-ready | |
CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
Mouse myeloma cell line NS0-derived recombinant human DNAM-1/CD226, Glu19-Asn247 (predicted), Accession # Q15762 | |
0.1 mL | |
Cellular Markers, Innate Immunity, Signal Transduction | |
10666 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction