Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAM-1/CD226 Antibody (26), FITC

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290489F
Description
DNAM-1/CD226 Monoclonal specifically detects DNAM-1/CD226 in Mouse samples. It is validated for Flow Cytometry, ELISA.Specifications
DNAM-1/CD226 | |
Monoclonal | |
FITC | |
CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DNAM-1/CD226 (NP_848802.2; Met 1-Pro 254). | |
0.1 mL | |
Cellular Markers, Innate Immunity, Signal Transduction | |
10666 | |
Mouse | |
Purified |
Flow Cytometry, ELISA | |
26 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human CD226/ DNAM-1 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction