Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAM-1/CD226 Antibody (2770B), PE/Atto594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10988PEATT594
Description
DNAM-1/CD226 Monoclonal antibody specifically detects DNAM-1/CD226 in Human samples. It is validated for Flow CytometrySpecifications
DNAM-1/CD226 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2770B | |
Flow Cytometry | |
CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
Human embryonic kidney cell HEK293-derived human DNAM-1/CD226 protein, Glu19-Asn247, Accession # Q15762 | |
0.1 mL | |
Cellular Markers, Innate Immunity, Signal Transduction | |
10666 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction