Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DNase I Solution (2500 U/mL) PROMO

Catalog No. PI90083PM
Click to view available options
:
Thermo Scientific™ DNase I Solution (2500 U/mL)

Catalog No. PI90083PM

Supplier: Thermo Scientific™ 90083

Improve protein extraction efficiency with this glycosylated polypeptide that cleaves single- and double-stranded DNA in cell lysates.

Deoxyribonuclease I (DNase I) is a single, glycosylated polypeptide that degrades unwanted single- and double-stranded DNA. The enzyme works by cleaving DNA into 5' phosphodinucleotide and small oligonucleotide fragments. DNase I is commonly added to cell lysis reagents to remove the viscosity caused by the DNA content in bacterial cell lysates or to remove the DNA templates from RNAs produced by in vitro transcription. Two grades of DNAse are offered, one sufficient for protein work and one that is useful for any application requiring the digestion of DNA in which it is crucial to avoid damage to RNA.

Highlights:

  • Degrades and removes unwanted DNA from samples
  • Cleaves both single-stranded and double-stranded DNA
  • Compatible with Thermo Scientific Pierce Cell Lysis Reagents
  • Reduces viscosity of bacterial lysates (protein extracts) to facilitate pipetting
  • Supplied as 50% glycerol stocks in 10 mM Tris-HCl pH 7.5, 10 mM CaCl2, 10 mM MgCl2
  • Available in two convenient formats: RNase-tested to protect RNA (Part No. 89835) and convenient protein extraction grade (Part No. 90083)

Recommended for:

Elimination of viscosity caused by DNA in protein cell lysates; Removal of DNA templates from RNAs produced by in vitro transcription

Specifications

Product Type Cell Lysis Enzyme
Content And Storage Store at -20°C
Form Liquid
Enzyme DNase
Quantity 0.5 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.