Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNase II Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA130778
Description
Recommended positive controls: Human spleen tissue lysate. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DNase II is a member of the DNase family. The protein, located in the lysosome, hydrolyzes DNA under acidic conditions and mediates the breakdown of DNA during erythropoiesis and apoptosis. Two codominant alleles have been characterized, DNASE2*L (low activity) and DNASE2*H (high activity), that differ at one nucleotide in the promoter region. The DNASE2*H allele is represented in this record.
Specifications
DNase II | |
Polyclonal | |
Unconjugated | |
DNASE2 | |
acid DNase; deoxyribonuclease; Deoxyribonuclease II alpha; deoxyribonuclease II, lysosomal; deoxyribonuclease-2-alpha; DNase II alpha; DNase II, lysosomal; DNASE2; Dnase2a; DNL; Dnl2; lysosomal DNase II; R31240_2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1777 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.4 | |
O00115 | |
DNASE2 | |
Peptide corresponding to aa 347- 360 of human DNase II precursor (accession no. AF047016). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction