Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNase II Polyclonal Antibody
DNase II Polyclonal Antibody
Supplier: Thermo Scientific PA584716
Description
DNase II Polyclonal Antibody for IHC (P)

Specifications
DNase II | |
Polyclonal | |
Unconjugated | |
DNASE2 | |
acid DNase; deoxyribonuclease; Deoxyribonuclease II alpha; deoxyribonuclease II, lysosomal; deoxyribonuclease-2-alpha; DNase II alpha; DNase II, lysosomal; DNASE2; Dnase2a; DNL; Dnl2; lysosomal DNase II; R31240_2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1777 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00115 | |
DNASE2 | |
Recombinant protein corresponding to Human DNASE2. Recombinant protein control fragment (Product #RP-103624). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction