Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNASE1 Monoclonal Antibody (2-1#), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543851
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543851 100 μL
PIMA543852 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543851 Supplier Invitrogen™ Supplier No. MA543851
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNASE1 Monoclonal antibody specifically detects DNASE1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNASE1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2-1#
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene DNASE1
Gene Accession No. P24855
Gene Alias deoxyribonuclease 1; deoxyribonuclease I; Deoxyribonuclease-1; DNase I; DNase I, lysosomal; DNASE1; DNL1; Dornase alfa; DRNI; epithelial lens deoxyribonuclease I; human urine deoxyribonuclease I; pancreatic deoxyribonuclease I
Gene Symbols DNASE1
Host Species Mouse
Immunogen Gly19-Ala259 (Accession P24855), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1773
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.