Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNASE1L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564477
Description
Immunogen sequence: GFHWVIADGE DTTVRASTHC TYDRVVLHGE RCRSLLHTAA AFDFPTSFQL TEEEALNISD H Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 75%.
DNASE1L1 encodes a deoxyribonuclease protein that shows high sequence similarity to DNase I. The encoded protein is localized to the endoplasmic reticulum and modified by N-linked glycosylation. Alternate transcriptional splice variants encoding the same protein have been observed.
Specifications
DNASE1L1 | |
Polyclonal | |
Unconjugated | |
DNASE1L1 | |
2310005K03Rik; AI324885; deoxyribonuclease; deoxyribonuclease 1 like 1; deoxyribonuclease 1-like 1; deoxyribonuclease I-like 1; Deoxyribonuclease-1-like 1; DNAS1L1; DNase I, lysosomal-like; DNase I-like 1; DNase I-like, muscle-specific; DNase X; Dnase1l1; Dnase1ll; Dnasex; DNL1L; Dnl1ll; G4.8; Muscle-specific DNase I-like; XIB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1774 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49184 | |
DNASE1L1 | |
Recombinant protein corresponding to Human DNASE1L1. Recombinant protein control fragment (Product #RP-103838). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction