Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNASE1L2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00
Specifications
Antigen | DNASE1L2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19142920
![]() |
Novus Biologicals
NBP19142920UL |
20 μL |
Each of 1 for $206.00
|
|
|||||
Description
DNASE1L2 Polyclonal specifically detects DNASE1L2 in Rat samples. It is validated for Western Blot.Specifications
DNASE1L2 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
deoxyribonuclease I-like 2, deoxyribonuclease-1-like 2, DHP1, DNAS1L2, DNase I homolog protein DHP1, DNase I-like 2 | |
DNASE1L2 | |
IgG | |
Affinity Purified | |
40 kDa |
Western Blot | |
Unconjugated | |
RUO | |
XP_002724704 | |
1775 | |
Synthetic peptide directed towards the middle region of human LOC100364462. Peptide sequence LIPLHAAPNQAVAEIDALYDVYLDVIDKWNTDDMLFLGDFNADCKYVKAH. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title