Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNASE1L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549535
Description
DNASE1L2 Polyclonal Antibody for Western Blot
Divalent cation-dependent acid DNA endonuclease involved in the breakdown of the nucleus during corneocyte formation of epidermal keratinocytes. May play an immune role by eliminating harmful DNA released into the extracellular environment by damaged epidermal cells.
Specifications
DNASE1L2 | |
Polyclonal | |
Unconjugated | |
DNASE1L2 | |
4733401H14Rik; deoxyribonuclease 1-like 2; deoxyribonuclease I-like 2; Deoxyribonuclease-1-like 2; DHP1; DNAS1L2; DNase I homolog protein DHP1; DNase I-like 2; Dnase1l2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1775 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q92874 | |
DNASE1L2 | |
KLH conjugated synthetic peptide between 199-229 amino acids from human DNASE1L2. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction