Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNASE1L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568413
Description
DNASE1L2 Polyclonal Antibody for IHC (P)
Divalent cation-dependent acid DNA endonuclease involved in the breakdown of the nucleus during corneocyte formation of epidermal keratinocytes. May play an immune role by eliminating harmful DNA released into the extracellular environment by damaged epidermal cells.
Specifications
DNASE1L2 | |
Polyclonal | |
Unconjugated | |
DNASE1L2 | |
4733401H14Rik; deoxyribonuclease 1-like 2; deoxyribonuclease I-like 2; Deoxyribonuclease-1-like 2; DHP1; DNAS1L2; DNase I homolog protein DHP1; DNase I-like 2; Dnase1l2 | |
Rabbit | |
affinity chromatography | |
RUO | |
1775 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q92874 | |
DNASE1L2 | |
Synthesized peptide derived from human DNASE1L2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction