Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNASE1L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107113
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107113 Supplier Invitrogen™ Supplier No. PA5107113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNASE1L3 Polyclonal Antibody for Western Blot

This gene encodes a member of the DNase family. The protein hydrolyzes DNA, is not inhibited by actin, and mediates the breakdown of DNA during apoptosis. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNASE1L3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene DNASE1L3
Gene Accession No. Q13609
Gene Alias deoxyribonuclease 1-like 3; deoxyribonuclease gamma; deoxyribonuclease I like 3; deoxyribonuclease I-like 3; deoxyribonuclease I-like III; DHP2; DNAS1L3; DNase gamma; DNase I homolog protein 2; DNase I homolog protein DHP2; DNase I-like 3; DNASE1L3; DNasegamma; DNaseY; Liver and spleen DNase; LSD; LS-DNase; macrophage specific DNase I; SLEB16
Gene Symbols DNASE1L3
Host Species Rabbit
Immunogen Synthetic peptide located within the following region: DFQKAYKLTE EEALDVSDHF PVEFKLQSSR AFTNSKKSVT LRKKTKSKRS (aa 256-305).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1776
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.