Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNASE1L3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 113301AP150UL

Description
This gene encodes a member of the DNase family. The protein hydrolyzes DNA, is not inhibited by actin, and mediates the breakdown of DNA during apoptosis. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.Specifications
DNASE1L3 | |
Polyclonal | |
Unconjugated | |
DNASE1L3 | |
Deoxyribonuclease gamma, deoxyribonuclease I like 3, DHP2, DNAS1L3, DNase gamma, DNase I homolog protein DHP2, DNase I like 3, DNASE1L3, Liver and spleen DNase, LS DNase, LSD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116687, 13421, 1776 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O55070, O89107, Q13609 | |
DNASE1L3 | |
DNASE1L3 Fusion Protein Ag1873 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction