Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNASE2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100482
Description
Antibody detects endogenous levels of total DNASE2B.
The protein encoded by this gene shares considerable sequence similarity to, and is structurally related to DNase II. The latter is a well characterized endonuclease that catalyzes DNA hydrolysis in the absence of divalent cations at acidic pH. Unlike DNase II which is ubiquitously expressed, expression of this gene product is restricted to the salivary gland and lungs. The gene has been localized to chromosome 1p22.3 adjacent (and in opposite orientation) to the uricase pseudogene. Two transcript variants encoding different isoforms have been described for this gene.
Specifications
DNASE2B | |
Polyclonal | |
Unconjugated | |
DNASE2B | |
deoxyribonuclease DLAD; deoxyribonuclease II beta; Deoxyribonuclease-2-beta; DLAD; DNA for uricase; DNase II beta; DNase II-like acid DNase; Dnase2b; DNase2-like acid DNase; DNaseII-like acid DNase; endonuclease DLAD; lysosomal DNase II; UOX | |
Rabbit | |
Affinity chromatography | |
RUO | |
58511, 59296 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WZ79, Q9QZK9 | |
DNASE2B | |
A synthesized peptide derived from human DNASE2B(Accession Q8WZ79), corresponding to amino acid residues Y302-G352. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction