Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAzol™ Reagent, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
200 mL
Description
General description
- Cholesterol (cholesterin, cholesteryl alcohol) is a lipid sterol found in vertebrate cell membranes
- Cholesterol is a cholestanoid, a class of molecule that contains the C-27 cholestane motif
- A precursor of many steroids and sex hormones in animals including cortisol, aldosterone, testosterone, progesterone, and estrogen
- It plays a role in neurotransmission and is crucial for organ systems, such as immune, reproductive and endocrine systems
Applications
- Suitable for in vitro and in vivo applications
- Stabilized in powder form for laboratory use only

Specifications
Specifications
Color | Blue-green |
For Use With (Application) | Complete and ready to use reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin |
Product Type | DNA Extraction Reagent |
Quantity | 200 mL |
Form | Liquid |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction