Learn More
Invitrogen™ DND1 Monoclonal Antibody (1F7)

Description
Peptide Sequence: EARLLPSPGP APGQIALLKF SSHRAAAMAK KALVEGQSHL CGEQVAVEWL KPDLKQRLRQ QLVGPFLRSP QPEGSQLALA RDKLGFQGAR ATLQ.
Specifications
Specifications
| Antigen | DND1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1F7 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | DND1 |
| Gene Accession No. | Q8IYX4 |
| Gene Alias | BC034897; dead end homolog 1; dead end homolog 1 (zebrafish); dead end protein homolog 1; DND microRNA-mediated repression inhibitor 1; Dnd1; MGC34750; Rbms4; RNA-binding motif, single-stranded-interacting protein 4; Ter; teratoma |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.