Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DND1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983264
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3264 Supplier Bioss Supplier No. BS14397R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DND1 contains 2 RRM (RNA recognition motif) domains. It may play a role during primordial germ cell (PGC) development. However, DND1 does not seem to be essential for PGC migration.

Specifications

Antigen DND1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DND1
Gene Accession No. Q8IYX4
Gene Alias BC034897; BOS_8203; dead end homolog 1; dead end homolog 1 (zebrafish); Dead end protein homolog 1; DND microRNA-mediated repression inhibitor 1; Dnd1; LOC100347217; MGC34750; RBMS4; RNA-binding motif, single-stranded-interacting protein 4; Ter; teratoma
Gene Symbols DND1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DND1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 373863
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.