Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNER Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120373 Supplier Invitrogen™ Supplier No. PA5120373
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Hela, HT-29, Mouse brain, Mouse eye, Mouse small intestine, Rat brain. The target is usually found in the following locations: Cell membrane, Single-pass type I membrane protein. Immunogen sequence: PATGWTESMA PRQLQPVPAT QEPDKILPRS QATVTLPTWQ PKTGQKVVEM KWDQVEVIPD IACGNASSNS SAGGRLVSFE VPQNTSVKIR QDATASLILL W.

DNER (Delta-notch-like EGF-related receptor), also known as Delta-notch-like EGF repeat-containing transmembrane protein, is a neuron-specific, atypical Notch ligand expressed in dendrites and cell bodies of neurons throughout the central nervous system. DNER contains ten extracellular EGF-like domains that are highly homologous to those of the Notch ligand, Delta. In the cerebellum, DNER is predominantly expressed in Purkinje cells. DNER mediates neuron-glia interaction during astrocytogenesis through a direct interaction with Notch 1 at Purkinje cell/Bergmann glia contacts. This interaction activates a Deltex-dependent Notch signaling pathway in Bergmann glia and may regulate Bergmann glial morphogenesis. DNER is crucial for the functional and morphological maturation of Bergmann glia. DNER-knockout mice are characterized by motor discoordination and cerebellum retardation in morphogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNER
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Dner
Gene Accession No. Q8JZM4, Q8NFT8
Gene Alias A930026D19Rik; BET; Brain EGF repeat-containing transmembrane protein; Bret; Delta and Notch-like epidermal growth factor-related receptor; delta/notch like EGF repeat containing; delta/notch-like EGF repeat containing; delta/notch-like EGF-related receptor; delta-notch-like EGF repeat-containing transmembrane; Dner; H_NH0150O02.1; LOW QUALITY PROTEIN: delta and Notch-like epidermal growth factor-related receptor; transmembrane protein Bet; UNQ26; UNQ262/PRO299; WUGSC:H_NH0150O02.1
Gene Symbols Dner
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227325, 316573, 92737
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.