Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNER Polyclonal Antibody, Invitrogen™

Catalog No. PIPA543631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543631 Supplier Thermo Scientific Supplier No. PA543631

Rabbit Polyclonal Antibody

Peptide sequence: DACQRKPCQN NASCIDANEK QDGSNFTCVC LPGYTGELCQ SKIDYCILDP Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 86%; Rabbit: 93%; Rat: 86%

DNER Polyclonal antibody specifically detects DNER in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNER
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Dner
Gene Accession No. Q8NFT8
Gene Alias A930026D19Rik; BET; Brain EGF repeat-containing transmembrane protein; Bret; Delta and Notch-like epidermal growth factor-related receptor; delta/notch like EGF repeat containing; delta/notch-like EGF repeat containing; delta/notch-like EGF-related receptor; delta-notch-like EGF repeat-containing transmembrane; Dner; H_NH0150O02.1; LOW QUALITY PROTEIN: delta and Notch-like epidermal growth factor-related receptor; transmembrane protein Bet; UNQ26; UNQ262/PRO299; WUGSC:H_NH0150O02.1
Gene Symbols Dner
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DNER
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92737
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.