Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNER Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206775
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6775 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6775 Supplier Fabgennix Inc Supplier No. DNER101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNER Polyclonal antibody specifically detects DNER in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen DNER
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Dner
Gene Accession No. Q8JZM4, Q8NFT8
Gene Alias A930026D19Rik; BET; Brain EGF repeat-containing transmembrane protein; Bret; Delta and Notch-like epidermal growth factor-related receptor; delta/notch like EGF repeat containing; delta/notch-like EGF repeat containing; delta/notch-like EGF-related receptor; delta-notch-like EGF repeat-containing transmembrane; Dner; H_NH0150O02.1; LOW QUALITY PROTEIN: delta and Notch-like epidermal growth factor-related receptor; transmembrane protein Bet; UNQ26; UNQ262/PRO299; WUGSC:H_NH0150O02.1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 700-737 on human Delta and Notch-like Epidermal growth factor-related receptorprotein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227325, 316573, 92737
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.