Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584355 Supplier Invitrogen™ Supplier No. PA584355
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CWHSLFKIQN RLAAMEDTST QGCQPVEITH LYPSGLSPQE FLGWLEGSCW HHGIFPKVLR AAGQCNNMGQ KRQTEESIGI QHWIICDGAS NGAWLD Highest antigen sequence indentity to the following orthologs: Mouse - 65%, Rat - 65%.

DNHD1 is a protein coding gene. Gene ontology (GO) annotation include dynein complex; extracellular exosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNHD1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNHD1
Gene Accession No. Q96M86
Gene Alias 8030491N06Rik; C11orf47; CCDC35; coiled-coil domain containing 35; DHCD1; DKFZp686J0796; DNHD1; DNHD1 variant protein; DNHD1L; dynein heavy chain domain 1; dynein heavy chain domain 1-like protein; Dynein heavy chain domain-containing protein 1; FLJ32752; FLJ35709; FLJ46184; Protein CCDC35; UNQ5781/PRO12970
Gene Symbols DNHD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNHD1. Recombinant protein control fragment (Product #RP-106792).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144132
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.