Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dNK (Deoxynucleoside Kinase) Polyclonal Antibody, Abbomax

Catalog No. 502204001
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4001 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4001 Supplier Abbomax Inc Supplier No. 601680
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Fruit-fly extract

dNK (Deoxynucleoside Kinase) Polyclonal antibody specifically detects dNK (Deoxynucleoside Kinase) in Chemical samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen dNK (Deoxynucleoside Kinase)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Alias DNK
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of fruit fly Deoxynucleoside kinase protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.