Learn More
DNL3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301637AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA Ligase III gene product migrates as a 100kDa (a-form) protein in somatic tissues, is ubiquitously expressed and complexes with XRCC-1 and a 97kDa (β-form) protein that is found in testis. DNA Ligase III is involved in rejoining DNA single-strand breaks and in the base excision repair pathway.Specifications
| DNL3 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| P49916 | |
| LIG3 | |
| Between 800 and 850 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| LIG3 | |
| LIG2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3980 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.