Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNLZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566527
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566527 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566527 Supplier Invitrogen™ Supplier No. PA566527
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGWFSDLNGKR NIEEILTARG EQVHRVAGEG ALELVLEAAG APTSTAAPEA GE Highest antigen sequence identity to the following orthologs - mouse 64%, rat 70%.

May function as a co-chaperone towards HSPA9/mortalin which, by itself, is prone to self-aggregation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNLZ
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNLZ
Gene Accession No. Q5SXM8
Gene Alias bA413M3.2; C9orf151; DNL-type zinc finger; DNL-type zinc finger protein; DNLZ; HEP; HEP1; HSP70 escort protein; hsp70-escort protein 1; mtHsp70-escort protein; RP11-413M3.2; TIMM15; translocase of inner mitochondrial membrane 15 homolog; ZIM17
Gene Symbols DNLZ
Host Species Rabbit
Immunogen Recombinant Human DNLZ. Recombinant protein control fragment (Product #RP-105240).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728489
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.