Learn More
DNM1L Mouse anti-Human, Clone: OTI3F3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506114

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
A human dynamin-related protein, DRP1 contributes to mitochondrial division in mammalian cells. It plays this important role in mitochondrial fission at steady state and during apoptosis. DRP1 is required for proper cellular distribution of mitochondria, and in mutant neurons, mitochondria are largely absent from synapses, thus providing a genetic tool to assess the role of mitochondria at synapses.Specifications
DNM1L | |
Monoclonal | |
Unconjugated | |
DNM1L | |
DLP1, DRP1, DVLP, DYMPLE, EMPF, EMPF1, HDYNIV, OPA5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10059 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3F3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O00429 | |
DNM1L | |
Full length human recombit protein of human DNM1L produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.