Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNM1L Mouse anti-Human, Mouse, Rat, Clone:4A4C7, Abnova™
Mouse monoclonal antibody raised against recombinant human DNM1L.
Supplier: Abnova Corporation MAB17781
Description
The protein encoded by this gene is a member of the dynamin superfamily of GTPases. Members of the dynamin-related subfamily, including the S. cerevisiae proteins Dnm1 and Vps1, contain the N-terminal tripartite GTPase domain but do not have the pleckstrin homology or proline-rich domains. This protein establishes mitochondrial morphology through a role in distributing mitochondrial tubules throughout the cytoplasm. The gene has 3 alternatively spliced transcripts encoding different isoforms. These transcripts are alternatively polyadenylated. [provided by RefSeq]Specifications
DNM1L | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunocytochemistry (1:200-1:1000) Immunohistochemistry Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
DNM1L | |
Mouse | |
100 μg | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
4A4C7 | |
dynamin 1-like | |
In ascites (0.03% sodium azide) | |
DLP1, DRP1, DVLP, DYMPLE, DYNIV-11, FLJ41912, HDYNIV, VPS1 | |
Recombinant protein corresponding to amino acids 69-213 of human DNM1L from E. coli. | |
10059 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction