Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNM1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597675
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597675 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597675 Supplier Invitrogen™ Supplier No. PA597675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNM1L Polyclonal Antibody for ELISA, IHC, IHC (P), Western Blot

A human dynamin-related protein, DRP1 contributes to mitochondrial division in mammalian cells. It plays this important role in mitochondrial fission at steady state and during apoptosis. DRP1 is required for proper cellular distribution of mitochondria, and in mutant neurons, mitochondria are largely absent from synapses, thus providing a genetic tool to assess the role of mitochondria at synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNM1L
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.535 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DNM1L
Gene Accession No. O00429, Q8K1M6
Gene Alias 6330417M19Rik; AI450666; C-terminal region; Dlp1; dnm1l; Dnm1p/Vps1p-like protein; Dnml1; Dnmlp1; Drp1; DVLP; DYMPLE; dynamin 1 like; dynamin 1-like; dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-1-like protein-like protein; Dynamin-like protein; dynamin-like protein 1; dynamin-like protein 4; Dynamin-like protein IV; dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; EGK_03514; EMPF; FLJ41912; HDYNIV; N-terminal region; python; similar to dynamin 1-like; VPS1; zgc:66163
Gene Symbols DNM1L
Host Species Rabbit
Immunogen Recombinant Human Dynamin-1-like protein (496-710AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10059, 74006
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.