Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNM1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-140 Supplier Invitrogen™ Supplier No. PA116987
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine based on 100% sequence homology. Suggested positive control: antigen standard for DNM1L (transient overexpression lysate), human kidney protein.

A human dynamin-related protein, DRP1 contributes to mitochondrial division in mammalian cells. It plays this important role in mitochondrial fission at steady state and during apoptosis. DRP1 is required for proper cellular distribution of mitochondria, and in mutant neurons, mitochondria are largely absent from synapses, thus providing a genetic tool to assess the role of mitochondria at synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNM1L
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DNM1L
Gene Accession No. O00429, O35303, Q8K1M6
Gene Alias 6330417M19Rik; AI450666; C-terminal region; Dlp1; dnm1l; Dnm1p/Vps1p-like protein; Dnml1; Dnmlp1; Drp1; DVLP; DYMPLE; dynamin 1 like; dynamin 1-like; dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-1-like protein-like protein; Dynamin-like protein; dynamin-like protein 1; dynamin-like protein 4; Dynamin-like protein IV; dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; EGK_03514; EMPF; FLJ41912; HDYNIV; N-terminal region; python; similar to dynamin 1-like; VPS1; zgc:66163
Gene Symbols DNM1L
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region within residues 500-600 of the human DRP1 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10059, 114114, 74006
Target Species Human, Mouse, Rat, Monkey, Fish
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.