Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNM2 Mouse anti-Human, Clone:3F5F3, Abnova™

Catalog No. 89334316 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-316 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-316 Supplier Abnova Corporation Supplier No. MAB17963
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human DNM2.

Dynamins represent one of the subfamilies of GTP-binding proteins. These proteins share considerable sequence similarity over the N-terminal portion of the molecule, which contains the GTPase domain. Dynamins are associated with microtubules. They have been implicated in cell processes such as endocytosis and cell motility, and in alterations of the membrane that accompany certain activities such as bone resorption by osteoclasts. Dynamins bind many proteins that bind actin and other cytoskeletal proteins. Dynamins can also self-assemble, a process that stimulates GTPase activity. Four alternatively spliced transcripts encoding different proteins have been described. Additional alternatively spliced transcripts may exist, but their full-length nature has not been determined. [provided by RefSeq]

Specifications

Antigen DNM2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3F5F3
Conjugate Unconjugated
Description dynamin 2
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Western Blot (1:200-1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene DNM2
Gene Alias CMTDI1, CMTDIB, DI-CMTB, DYN2, DYNII
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 520-744 of human DNM2.
Quantity 100 μg
Gene ID (Entrez) 1785
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.