Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNMBP Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171830AF488
Description
DNMBP Polyclonal antibody specifically detects DNMBP in Human samples. It is validated for Western BlotSpecifications
DNMBP | |
Polyclonal | |
Western Blot | |
dynamin binding protein, dynamin-binding protein, KIAA1010ARHGEF36, Scaffold protein Tuba, TUBA | |
The immunogen for this product maps to a region between residue 75 and 125 of human Scaffold Protein TUBA using the numbering given in entry NP_056036.1 (GeneID 23268). | |
0.1 mL | |
Signal Transduction | |
23268 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction