Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNMBP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171831
Description
DNMBP Polyclonal specifically detects DNMBP in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
DNMBP | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
dynamin binding protein, dynamin-binding protein, KIAA1010ARHGEF36, Scaffold protein Tuba, TUBA | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 mcg/mg | |
NP_056036.1 | |
DNMBP | |
The immunogen for this product maps to a region between residue 575 and 625 of human Scaffold Protein TUBA using the numbering given in entry NP_056036.1 (GeneID 23268). | |
0.1 mL | |
Signal Transduction | |
23268 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction