Learn More
Invitrogen™ DNMBP Monoclonal Antibody (1H2)

Description
Peptide Sequence: TKKPFERKTI DRQSARKPLL GLPSYMLQSE ELRASLLARY PPEKLFQAER NFNAAQDLDV SLLEGDLVGV IKKKDPMGSQ NRWLIDNGVT KGFVYSSFLK.
Specifications
Specifications
| Antigen | DNMBP |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1H2 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | DNMBP |
| Gene Accession No. | Q6XZF7 |
| Gene Alias | 2410003L07Rik; 2410003M15Rik; ARHGEF36; Dnmbp; dynamin binding protein; dynamin-binding protein; Kiaa1010; Scaffold protein Tuba; TUBA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.