Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1, Ac-4K Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABE195625UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1956-25UG 25 μg
ABE1956-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE1956-25UG Supplier MilliporeSigma Supplier No. ABE195625UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-DNMT1, Ac-4K, Cat. No. ABE1956, is a highly specific rabbit polyclonal antibody that targets DNA (cytosine-5)-methyltransferase 1 (DNMT1) and has been tested for use in Dot Blot, Immunocytochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNMT1, Ac-4K
Applications Dot Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.02% sodium azide and 50% glycerol.
Gene Alias DNA (cytosine-5)-methyltransferase 1;EC: 2.1.1.37;CXXC-type zinc finger protein 9;DNA methyltransferase HsaI;DNA MTase HsaI;M.HsaI;MCMT
Gene Symbols DNMT1;AIM;CXXC9;DNMT
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 11 amino acids from the C-terminal half with acetylated K1111, 1113, 1115, and 1117.
Molecular Weight of Antigen ∽200 kDa observed; 183.16 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001370
Test Specificity This rabbit polyclonal antibody specifically detects DNA (cytosine-5)-methyltransferase 1 acetylated on four lysine residues from the C-terminal half containing six tandem repeats of K-G.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.