Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNMT1 Antibody (DNMT1/2061), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308482B
Description
DNMT1 Monoclonal antibody specifically detects DNMT1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
DNMT1 | |
Monoclonal | |
Biotin | |
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992 | |
Recombinant human DNMT1 protein fragment (around aa 767-912) (exact sequence is proprietary) (Uniprot: P26358) | |
100 μg | |
Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Stem Cell Markers, Transcription Factors and Regulators | |
1786 | |
Store at 4°C in the dark. | |
IgG2b κ |
Immunohistochemistry (Paraffin) | |
DNMT1/2061 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction